Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204783.1 | NP_990114.1 | 4 - 5 | 775 | 66 | |
GenBank mRNA | D31900.1 | - | 4 - 5 | 775 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
