Assay Details
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291731.1 | NP_001278660.1 | 4 - 5 | 424 | 75 | |
NM_001291734.1 | NP_001278663.1 | 4 - 5 | 424 | 75 | ||
NM_001291736.1 | NP_001278665.1 | 2 - 3 | 220 | 75 | ||
GenBank mRNA | BU230611.1 | - | 2 - 3 | 230 | 75 | |
BU351827.1 | - | 2 - 3 | 163 | 74 | ||
BX929541.1 | - | 4 - 5 | 371 | 75 | ||
CO635554.1 | - | 4 - 5 | 355 | 75 | ||
CR385888.1 | - | 4 - 5 | 358 | 75 | ||
CR407481.1 | - | 4 - 5 | 395 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
