Assay Details
Gene Symbol: SYNGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318453.1 | NP_001305382.1 | 3 - 4 | 534 | 123 | |
GenBank mRNA | AJ719536.1 | - | 3 - 4 | 534 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
