Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115081.2 | NP_001108553.2 | 2 - 3 | 967 | 87 | |
GenBank mRNA | BU202250.1 | - | 1 - 2 | 303 | 87 | |
BU302582.1 | - | 1 - 2 | 224 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
