Assay Details
Gene Symbol: RPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321599.1 | NP_001308528.1 | 2 - 3 | 121 | 68 | |
GenBank mRNA | BX933858.2 | - | 2 - 3 | 122 | 68 | |
CR390619.1 | - | 1 - 2 | 380 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
