Assay Details
Gene Symbol: IGFBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204353.1 | NP_989684.1 | 2 - 3 | 559 | 100 | |
GenBank mRNA | AY230206.1 | - | 2 - 3 | 559 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
