Assay Details
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205129.1 | NP_990460.1 | 1 - 2 | 113 | 75 | |
XM_025145278.1 | XP_025001046.1 | 1 - 2 | 128 | 75 | ||
GenBank mRNA | D13721.1 | - | 1 - 2 | 113 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_205129.1, XM_025145278.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
