Assay Details
Gene Symbol: RELA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205129.1 | NP_990460.1 | 4 - 5 | 459 | 100 | |
XM_025145278.1 | XP_025001046.1 | 4 - 5 | 477 | 100 | ||
GenBank mRNA | D13721.1 | - | 4 - 5 | 459 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
