Assay Details
Gene Symbol: ADCYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001291.2 | NP_001001291.2 | 3 - 4 | 362 | 100 | |
GenBank mRNA | AY956323.1 | - | 3 - 4 | 341 | 100 | |
BU351981.1 | - | 4 - 5 | 389 | 100 | ||
BX932280.2 | - | 4 - 5 | 395 | 100 | ||
BX932926.2 | - | 4 - 5 | 436 | 100 | ||
U71183.1 | - | 4 - 5 | 592 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
