Assay Details
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001614.1 | NP_001001614.1 | 1 - 2 | 91 | 66 | |
GenBank mRNA | AY605246.1 | - | 1 - 2 | 91 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001001614.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
