Assay Details
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001614.1 | NP_001001614.1 | 3 - 4 | 264 | 107 | |
GenBank mRNA | AY605246.1 | - | 3 - 4 | 264 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
