Assay Details
Gene Symbol: THPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001784.1 | NP_001001784.1 | 2 - 3 | 174 | 92 | |
XM_015291386.1 | XP_015146872.1 | 1 - 2 | 943 | 92 | ||
XM_015291387.2 | XP_015146873.1 | 3 - 4 | 2406 | 92 | ||
XM_015291388.2 | XP_015146874.1 | 3 - 4 | 2060 | 92 | ||
GenBank mRNA | AY613434.1 | - | 2 - 3 | 174 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
