Assay Details
Gene Symbol: NDUFV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199170.1 | NP_001186099.1 | 3 - 4 | 1107 | 108 | |
XM_015300381.2 | XP_015155867.1 | 3 - 4 | 1140 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
