Assay Details
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199594.1 | NP_001186523.1 | 1 - 2 | 136 | 90 | |
GenBank mRNA | AJ445824.1 | - | 1 - 2 | 145 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001199594.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
