Assay Details
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199594.1 | NP_001186523.1 | 3 - 4 | 1811 | 98 | |
XM_025150685.1 | XP_025006453.1 | 3 - 4 | 1799 | 98 | ||
XM_025150686.1 | XP_025006454.1 | 4 - 5 | 1766 | 98 | ||
GenBank mRNA | BU305109.1 | - | 1 - 2 | 534 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
