Assay Details
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199594.1 | NP_001186523.1 | 4 - 5 | 1967 | 163 | |
XM_025150685.1 | XP_025006453.1 | 4 - 5 | 1955 | 163 | ||
XM_025150686.1 | XP_025006454.1 | 5 - 6 | 1922 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
