Assay Details
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243222.1 | NP_001230151.1 | 4 - 5 | 700 | 87 | |
GenBank mRNA | AJ446140.1 | - | 4 - 5 | 641 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
