Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256162.1 | NP_001243091.1 | 1 - 2 | 122 | 134 | |
XM_015290247.2 | XP_015145733.1 | 2 - 3 | 260 | 134 | ||
XM_015290254.2 | XP_015145740.1 | 2 - 3 | 411 | 134 | ||
XM_015290265.2 | XP_015145751.1 | 2 - 3 | 255 | 134 | ||
XM_015290272.2 | XP_015145758.1 | 2 - 3 | 263 | 134 | ||
XM_025148966.1 | XP_025004734.1 | 2 - 3 | 258 | 134 | ||
GenBank mRNA | HQ439437.1 | - | 1 - 2 | 122 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
