Assay Details
Gene Symbol: RAB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270662.1 | NP_001257591.1 | 2 - 3 | 145 | 129 | |
GenBank mRNA | BU129917.1 | - | 2 - 3 | 322 | 129 | |
CR353962.1 | - | 2 - 3 | 314 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
