Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277517.1 | NP_001264446.1 | 1 - 2 | 129 | 91 | |
GenBank mRNA | BX931195.1 | - | 1 - 2 | 129 | 91 | |
CR352559.1 | - | 1 - 2 | 133 | 91 | ||
CR406235.1 | - | 1 - 2 | 140 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
