Assay Details
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277570.1 | NP_001264499.1 | 3 - 4 | 499 | 111 | |
GenBank mRNA | BU450299.1 | - | 3 - 4 | 493 | 113 | |
BU459205.1 | - | 3 - 4 | 279 | 112 | ||
CO422157.1 | - | 3 - 4 | 545 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
