Assay Details
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277647.3 | NP_001264576.1 | 3 - 4 | 326 | 124 | |
NM_001277650.3 | NP_001264579.1 | 3 - 4 | 326 | 124 | ||
GenBank mRNA | BU253790.1 | - | 3 - 4 | 323 | 124 | |
BU381872.1 | - | 3 - 4 | 326 | 124 | ||
BX933762.2 | - | 3 - 4 | 312 | 124 | ||
BX935197.1 | - | 3 - 4 | 337 | 124 | ||
BX950699.2 | - | 2 - 3 | 349 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
