Assay Details
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277688.1 | NP_001264617.1 | 1 - 2 | 45 | 100 | |
NM_001277689.1 | NP_001264618.1 | 1 - 2 | 44 | 100 | ||
NM_001277690.1 | NP_001264619.1 | 1 - 2 | 45 | 100 | ||
GenBank mRNA | BM426266.1 | - | 1 - 2 | 45 | 100 | |
BX934045.2 | - | 1 - 2 | 45 | 100 | ||
CN236286.1 | - | 1 - 2 | 44 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277688.1, NM_001277689.1, NM_001277690.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
