Assay Details
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277791.1 | NP_001264720.1 | 5 - 6 | 565 | 96 | |
NM_001277792.1 | NP_001264721.1 | 5 - 6 | 440 | 96 | ||
NM_001277793.1 | NP_001264722.1 | 5 - 6 | 423 | 96 | ||
XM_015275581.2 | XP_015131067.1 | 5 - 6 | 472 | 96 | ||
XM_025150364.1 | XP_025006132.1 | 5 - 6 | 568 | 96 | ||
XM_025150365.1 | XP_025006133.1 | 5 - 6 | 621 | 96 | ||
XM_025150373.1 | XP_025006141.1 | 5 - 6 | 441 | 96 | ||
GenBank mRNA | BU338918.1 | - | 2 - 3 | 202 | 96 | |
BU340762.1 | - | 5 - 6 | 440 | 96 | ||
BX930968.2 | - | 5 - 6 | 423 | 96 | ||
BX934861.1 | - | 5 - 6 | 425 | 96 | ||
CR385455.1 | - | 5 - 6 | 440 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
