Assay Details
Gene Symbol: ESD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277791.1 | NP_001264720.1 | 6 - 7 | 683 | 104 | |
NM_001277792.1 | NP_001264721.1 | 6 - 7 | 558 | 104 | ||
NM_001277793.1 | NP_001264722.1 | 6 - 7 | 541 | 104 | ||
XM_015275581.2 | XP_015131067.1 | 6 - 7 | 590 | 104 | ||
XM_025150364.1 | XP_025006132.1 | 6 - 7 | 686 | 104 | ||
XM_025150365.1 | XP_025006133.1 | 6 - 7 | 739 | 104 | ||
XM_025150373.1 | XP_025006141.1 | 6 - 7 | 559 | 104 | ||
GenBank mRNA | BU338918.1 | - | 3 - 4 | 320 | 104 | |
BU340762.1 | - | 6 - 7 | 558 | 104 | ||
BX930968.2 | - | 6 - 7 | 541 | 104 | ||
BX934861.1 | - | 6 - 7 | 543 | 104 | ||
CR385455.1 | - | 6 - 7 | 558 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
