Assay Details
Gene Symbol: SDHAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277889.1 | NP_001264818.1 | 2 - 3 | 282 | 106 | |
NM_001277890.1 | NP_001264819.1 | 2 - 3 | 258 | 106 | ||
GenBank mRNA | BU205400.1 | - | 2 - 3 | 261 | 106 | |
BU353870.1 | - | 2 - 3 | 281 | 106 | ||
CN228764.1 | - | 2 - 3 | 237 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
