Assay Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277911.1 | NP_001264840.1 | 3 - 4 | 227 | 94 | |
XM_015284105.2 | XP_015139591.1 | 4 - 5 | 292 | 94 | ||
GenBank mRNA | AJ445022.1 | - | 3 - 4 | 227 | 94 | |
BU224688.1 | - | 4 - 5 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
