Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278042.1 | NP_001264971.1 | 2 - 3 | 380 | 120 | |
GenBank mRNA | BU250968.1 | - | 2 - 3 | 195 | 120 | |
BX930465.1 | - | 2 - 3 | 196 | 120 | ||
BX930887.2 | - | 2 - 3 | 223 | 120 | ||
BX933231.1 | - | 2 - 3 | 201 | 120 | ||
CO635559.1 | - | 3 - 4 | 492 | 120 | ||
CR352580.1 | - | 2 - 3 | 223 | 120 | ||
CR353677.1 | - | 1 - 2 | 547 | 120 | ||
CR407506.1 | - | 2 - 3 | 227 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
