Assay Details
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278101.3 | NP_001265030.1 | 2 - 3 | 520 | 74 | |
NM_001278103.3 | NP_001265032.1 | 2 - 3 | 218 | 74 | ||
GenBank mRNA | AJ451649.1 | - | 2 - 3 | 219 | 74 | |
BI066001.1 | - | 2 - 3 | 507 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001278103.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
