Assay Details
Gene Symbol: C1H12ORF57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278146.1 | NP_001265075.1 | 2 - 3 | 567 | 96 | |
XM_015293555.2 | XP_015149041.1 | 2 - 3 | 323 | 96 | ||
XM_015293562.1 | XP_015149048.1 | 2 - 3 | 255 | 96 | ||
GenBank mRNA | BU468243.1 | - | 2 - 3 | 272 | 96 | |
BX929337.2 | - | 2 - 3 | 369 | 96 | ||
BX931286.1 | - | 2 - 3 | 376 | 96 | ||
BX935168.2 | - | 2 - 3 | 569 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
