Assay Details
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293094.1 | NP_001280023.1 | 2 - 3 | 320 | 172 | |
GenBank mRNA | DT657687.1 | - | 2 - 3 | 332 | 172 | |
X91011.1 | - | 303 | 172 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
