Assay Details
Gene Symbol: CDC42SE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302117.1 | NP_001289046.1 | 2 - 3 | 420 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302117.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
