Assay Details
Gene Symbol: NRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302155.1 | NP_001289084.1 | 1 - 2 | 161 | 113 | |
GenBank mRNA | CN218451.1 | - | 1 - 2 | 161 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302155.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
