Assay Details
Gene Symbol: DUSP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318409.1 | NP_001305338.1 | 1 - 2 | 571 | 69 | |
XM_015292746.2 | XP_015148232.1 | 1 - 2 | 603 | 69 | ||
GenBank mRNA | AJ851815.1 | - | 1 - 2 | 571 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
