Assay Details
Gene Symbol: GNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015283546.2 | XP_015139032.1 | 4 - 5 | 664 | 60 | |
XM_015283547.2 | XP_015139033.1 | 3 - 4 | 550 | 60 | ||
GenBank mRNA | BX929533.1 | - | 4 - 5 | 540 | 60 | |
BX931782.2 | - | 3 - 4 | 501 | 60 | ||
BX931798.1 | - | 3 - 4 | 501 | 60 | ||
BX934428.1 | - | 4 - 5 | 597 | 60 | ||
BX935144.2 | - | 4 - 5 | 628 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
