Assay Details
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004937750.3 | XP_004937807.3 | 4 | 2521 | 83 | |
XM_004937751.3 | XP_004937808.3 | 3 | 2331 | 83 | ||
XM_425481.5 | XP_425481.4 | 4 | 2422 | 83 | ||
GenBank mRNA | CR353594.1 | - | 1 | 722 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
