Assay Details
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231624.5 | XP_001231625.2 | 78 | 92 | ||
GenBank mRNA | CR385970.1 | - | 80 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001231624.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
