Assay Details
Gene Symbol: BCL2L15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001233930.5 | XP_001233931.1 | 93 | |||
GenBank mRNA | CR388427.1 | - | 605 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
