Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004946646.3 | XP_004946703.1 | 7 - 8 | 864 | 85 | |
XM_015295831.2 | XP_015151317.2 | 7 - 8 | 1051 | 85 | ||
XM_015295832.2 | XP_015151318.1 | 6 - 7 | 1003 | 85 | ||
XM_015295834.2 | XP_015151320.1 | 2 - 3 | 202 | 85 | ||
GenBank mRNA | CR390232.1 | - | 6 - 7 | 625 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
