Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231860.5 | XP_001231861.1 | 2 - 3 | 204 | 125 | |
GenBank mRNA | BX930182.2 | - | 2 - 3 | 161 | 125 | |
BX930637.2 | - | 2 - 3 | 181 | 125 | ||
BX930766.1 | - | 2 - 3 | 153 | 125 | ||
BX932061.1 | - | 2 - 3 | 191 | 125 | ||
BX932780.2 | - | 2 - 3 | 151 | 125 | ||
BX934576.2 | - | 2 - 3 | 201 | 125 | ||
BX934964.2 | - | 2 - 3 | 159 | 125 | ||
BX950797.1 | - | 2 - 3 | 194 | 125 | ||
CR385887.1 | - | 2 - 3 | 181 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
