Assay Details
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003641013.4 | XP_003641061.1 | 2 - 3 | 252 | 67 | |
GenBank mRNA | BX930224.1 | - | 2 - 3 | 241 | 67 | |
BX934451.1 | - | 2 - 3 | 251 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
