Assay Details
Gene Symbol: MYCT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419680.6 | XP_419680.1 | 1 - 2 | 148 | 69 | |
GenBank mRNA | CR524378.1 | - | 1 - 2 | 117 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
