Assay Details
Gene Symbol: LOC107057310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025142368.1 | XP_024998136.1 | 15 - 16 | 1052 | 84 | |
XM_025142369.1 | XP_024998137.1 | 15 - 16 | 1072 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
