Assay Details
Gene Symbol: LOC107057310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025142366.1 | XP_024998134.1 | 3 - 4 | 191 | 65 | |
XM_025142367.1 | XP_024998135.1 | 3 - 4 | 264 | 65 | ||
XM_025142368.1 | XP_024998136.1 | 3 - 4 | 191 | 65 | ||
XM_025142369.1 | XP_024998137.1 | 3 - 4 | 211 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
