Assay Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293044.2 | XP_015148530.1 | 4 - 5 | 675 | 96 | |
XM_416489.6 | XP_416489.2 | 3 - 4 | 425 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
