Assay Details
Gene Symbol: ZC3HAV1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293044.2 | XP_015148530.1 | 6 - 7 | 820 | 124 | |
XM_025142641.1 | XP_024998409.1 | 4 - 5 | 401 | 124 | ||
XM_416489.6 | XP_416489.2 | 5 - 6 | 570 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
