Assay Details
Gene Symbol: VHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025154544.1 | XP_025010312.1 | 2 | 3088 | 100 | |
XM_414447.5 | XP_414447.3 | 3 | 2128 | 100 |
Gene Symbol: IRAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293126.2 | XP_015148612.1 | 1 | 1469 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015293126.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
