Assay Details
Gene Symbol: C1QTNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004941560.3 | XP_004941617.3 | 1 - 2 | 107 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004941560.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
