Assay Details
Gene Symbol: USP51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294380.2 | XP_015149866.1 | 10 - 11 | 5801 | 63 | |
XM_015294382.2 | XP_015149868.2 | 6 - 7 | 667 | 63 | ||
XM_015294383.2 | XP_015149869.1 | 11 - 12 | 5846 | 63 | ||
XM_025155364.1 | XP_025011132.1 | 11 - 12 | 6872 | 63 | ||
XM_025155365.1 | XP_025011133.1 | 8 - 9 | 949 | 63 | ||
XM_025155366.1 | XP_025011134.1 | 9 - 10 | 5657 | 63 | ||
XM_414805.6 | XP_414805.3 | 7 - 8 | 781 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
