Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299058.2 | XP_015154544.1 | 6433 | 77 | ||
XM_015299059.2 | XP_015154545.1 | 5960 | 77 | |||
XM_015299060.2 | XP_015154546.1 | 6028 | 77 | |||
XM_015299061.2 | XP_015154547.1 | 6210 | 77 | |||
XM_025143773.1 | XP_024999541.1 | 6848 | 77 |
Gene Symbol: MAPKAPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015299062.2 | XP_015154548.1 | 2179 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015299062.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
